Gene Transcripts

Gene Symbol
Pcdhgc4
Entrez Gene ID
93707
Gene Name
protocadherin gamma subfamily C, 4
Gene Aliases
Chromosome Location
Chr. 18: 37815079 - 37841873 on Build GRCm38
UniGene ID
Mm.247203
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033582.2 NP_291060.1 1 591
GenBank mRNA AK139465.1 - 1 215
AY013812.1 - 1 137
BC064808.1 - 1 591 6825007

View Full Product Details