Gene Transcripts

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
Chromosome Location
Chr. 21: 36750888 - 36990254 on Build GRCh38
UniGene ID
Hs.371350
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000411.6 NP_000402.3 12 5644
NM_001242784.1 NP_001229713.1 12 6578
NM_001242785.1 NP_001229714.1 12 5737
XM_005260953.3 XP_005261010.1 11 5606
XM_005260955.3 XP_005261012.1 11 5477
XM_005260956.3 XP_005261013.1 11 5653
XM_006723994.2 XP_006724057.1 11 5591
XM_011529541.2 XP_011527843.1 11 5480
XM_017028330.1 XP_016883819.1 11 5427
GenBank mRNA BI094308.1 - 1 197
D87328.1 - 12 6394

View Full Product Details