Gene Transcripts
- Gene Symbol
- Egln2
- Entrez Gene ID
- 112406
- Gene Name
- egl-9 family hypoxia-inducible factor 2
- Gene Aliases
- 0610011A13Rik, C85656, Hif-p4h-1, Ier4, Phd1, SM-20
- Chromosome Location
- Chr. 7: 27158658 - 27166802 on Build GRCm38
- UniGene ID
- Mm.29978
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053208.4 |
NP_444438.2
|
2, 3 |
1236 |
|
GenBank mRNA |
AF340231.1 |
- |
1, 2 |
978 |
|
AF453879.1 |
- |
2, 3 |
1158 |
|
AJ310547.1 |
- |
2, 3 |
1207 |
|
AK075582.1 |
- |
2, 3 |
1188 |
|
AK152056.1 |
- |
2, 3 |
1197 |
|
AK152223.1 |
- |
2, 3 |
1194 |
|
AK159206.1 |
- |
2, 3 |
1189 |
|
AK166661.1 |
- |
3, 4 |
1102 |
|
AK167477.1 |
- |
2, 3 |
1189 |
|
BC023299.1 |
- |
2, 3 |
1170 |
5135579 |
BC051436.1 |
- |
1, 2 |
954 |
6312638 |
BC086764.1 |
- |
2, 3 |
1175 |
5685311 |
View Full Product Details