Gene Transcripts

Gene Symbol
Egln2
Entrez Gene ID
112406
Gene Name
egl-9 family hypoxia-inducible factor 2
Gene Aliases
0610011A13Rik, C85656, Hif-p4h-1, Ier4, Phd1, SM-20
Chromosome Location
Chr. 7: 27158658 - 27166802 on Build GRCm38
UniGene ID
Mm.29978
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053208.4 NP_444438.2 6 1576
GenBank mRNA AF340231.1 - 5 1318
AF453879.1 - 6 1498
AJ310547.1 - 6 1547
AK075582.1 - 6 1528
AK152056.1 - 6 1537
AK152223.1 - 6 1534
AK159206.1 - 6 1529
AK166661.1 - 7 1442
AK167477.1 - 6 1529
AK179611.1 - 3 158
BC023299.1 - 6 1510 5135579
BC051436.1 - 5 1294 6312638
BC086764.1 - 6 1515 5685311

View Full Product Details