Gene Transcripts

Gene Symbol
Foxp1
Entrez Gene ID
108655
Gene Name
forkhead box P1
Gene Aliases
3110052D19Rik, 4932443N09Rik, AI461938, AW494214
Chromosome Location
Chr. 6: 98925342 - 99523024 on Build GRCm38
UniGene ID
Mm.234965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197321.1 NP_001184250.1 7 858
NM_001197322.1 NP_001184251.1 4 465
NM_053202.2 NP_444432.1 8 1010
GenBank mRNA AB221601.1 - 5 705
AB221602.1 - 5 703
AB594192.1 - 9 1418
AF339103.1 - 5 699
AF339104.1 - 5 699
AK033368.1 - 2 175
AK077062.1 - 4 360
AK139831.1 - 4 351
BC064764.1 - 7 844 30430449

View Full Product Details