Gene Transcripts

Gene Symbol
SFTPB
Entrez Gene ID
6439
Gene Name
surfactant protein B
Gene Aliases
PSP-B, SFTB3, SFTP3, SMDP1, SP-B
Chromosome Location
Chr. 2: 85657317 - 85668741 on Build GRCh38
UniGene ID
Hs.512690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000542.3 NP_000533.3 4 363
NM_198843.2 NP_942140.2 4 363
XM_005264487.2 XP_005264544.1 3 278
XM_005264488.3 XP_005264545.2 3 243
XM_005264490.4 XP_005264547.2 3 243
XM_017004702.1 XP_016860191.1 3 278
XM_017004703.1 XP_016860192.1 3 243
GenBank mRNA AK314947.1 - 3 243
AL548140.3 - 4 363
BC032785.1 - 3 230 5179408
BI821350.1 - 233
DQ317589.1 - 3 216
J02761.1 - 3 240

View Full Product Details