Gene Transcripts

Gene Symbol
Xpnpep2
Entrez Gene ID
170745
Gene Name
X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound
Gene Aliases
9030008G12Rik, mAPP
Chromosome Location
Chr. X: 48106913 - 48136982 on Build GRCm38
UniGene ID
Mm.129279
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289729.1 NP_001276658.1 2 518
NM_133213.2 NP_573476.2 2 518
GenBank mRNA AF367247.1 - 2 182
AF428102.1 - 2 369
AK078835.1 - 2 325
AK087627.1 - 2 325
AK164538.1 - 2 346
AK172086.1 - 2 519
BC140977.1 - 2 162 9055589

View Full Product Details