Gene Transcripts

Gene Symbol
Krt75
Entrez Gene ID
109052
Gene Name
keratin 75
Gene Aliases
4732468K03Rik, AA589387, K6hf, Krt2-6hf, Krtcap1
Chromosome Location
Chr. 15: 101563343 - 101573904 on Build GRCm38
UniGene ID
Mm.106868
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133357.3 NP_579935.1 1 227
GenBank mRNA AK028972.1 - 1 228
AK037233.1 - 1 228
AK076367.1 - 1 228
BC137935.1 - 1 228 8860954
BC137936.1 - 1 228 8860955

View Full Product Details