Gene Transcripts

Gene Symbol
Ttc1
Entrez Gene ID
66827
Gene Name
tetratricopeptide repeat domain 1
Gene Aliases
4833412C19Rik, C79328, TPR1
Chromosome Location
Chr. 11: 43730006 - 43747973 on Build GRCm38
UniGene ID
Mm.271974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133795.1 NP_598556.1 2 255
GenBank mRNA AK019021.1 - 2 251
AK077715.1 - 2 268
AK166431.1 - 2 262
AK170032.1 - 2 265
AK170230.1 - 2 265
BC010236.1 - 2 255 3581904

View Full Product Details