Gene Transcripts

Gene Symbol
Psph
Entrez Gene ID
100678
Gene Name
phosphoserine phosphatase
Gene Aliases
AI480570, PSP, PSPase
Chromosome Location
Chr. 5: 129765558 - 129787253 on Build GRCm38
UniGene ID
Mm.271784
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133900.4 NP_598661.1 4 722
GenBank mRNA AK088865.1 - 4 674
AK168048.1 - 2 330
BC002251.1 - 2 293 3493291
BQ177662.1 - 2 288

View Full Product Details