Gene Transcripts

Gene Symbol
Cisd1
Entrez Gene ID
52637
Gene Name
CDGSH iron sulfur domain 1
Gene Aliases
AU043990, AW743335, D10Ertd214e, Zcd1, mitoNEET
Chromosome Location
Chr. 10: 71330494 - 71344849 on Build GRCm38
UniGene ID
Mm.254114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134007.4 NP_598768.1 3 421
GenBank mRNA AK075907.1 - 3 422
AK131595.1 - 3 421
AK131605.1 - 3 400
AK131812.1 - 3 425
AK131995.1 - 3 432
AK167898.1 - 3 400
BC013522.1 - 3 410 4236502
BC019860.1 - 3 437 5123692
BC021952.1 - 3 458 5102315

View Full Product Details