Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083902.1 | NP_001077371.1 | 8 | 1333 | |
NM_134251.2 | NP_599012.2 | 9 | 1449 | ||
GenBank mRNA | AF319951.1 | - | 9 | 1443 | |
AK087781.1 | - | 5 | 1039 | ||
AK165080.1 | - | 4 | 956 | ||
BC030926.1 | - | 8 | 1333 | 4910321 |