Gene Transcripts
- Gene Symbol
- Cds2
- Entrez Gene ID
- 110911
- Gene Name
- CDP-diacylglycerol synthase (phosphatidate cytidylyltransferase) 2
- Gene Aliases
- 5730450N06Rik, 5730460C18Rik, AI854580, D2Wsu127e
- Chromosome Location
- Chr. 2: 132262609 - 132312050 on Build GRCm38
- UniGene ID
- Mm.284503
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001291039.1 |
NP_001277968.1
|
5, 6 |
507 |
|
NM_138651.7 |
NP_619592.1
|
5, 6 |
830 |
|
GenBank mRNA |
AK036328.1 |
- |
5, 6 |
681 |
|
AK049257.1 |
- |
5, 6 |
508 |
|
AK147541.1 |
- |
5, 6 |
722 |
|
AK166001.1 |
- |
5, 6 |
681 |
|
AY159802.1 |
- |
5, 6 |
753 |
|
BC003852.1 |
- |
5, 6 |
736 |
3600825 |
BC069879.1 |
- |
5, 6 |
709 |
30439069 |
CD540005.1 |
- |
5, 6 |
505 |
|
View Full Product Details