Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144512.2 | NP_653095.1 | 7 | 828 | |
GenBank mRNA | AK078399.1 | - | 7 | 835 | |
AK149557.1 | - | 7 | 836 | ||
BC023117.1 | - | 7 | 816 | 4240641 | |
BC029637.1 | - | 7 | 828 | 4195373 |