Gene Transcripts

Gene Symbol
2210407C18Rik
Entrez Gene ID
78354
Gene Name
RIKEN cDNA 2210407C18 gene
Gene Aliases
Ep1
Chromosome Location
Chr. 11: 58608204 - 58613492 on Build GRCm38
UniGene ID
Mm.58038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144544.2 NP_653127.2 6 613
GenBank mRNA AA434919.1 - 4 419
AY148487.1 - 5 519
BC019553.1 - 6 591 4456658

View Full Product Details