Gene Transcripts

Gene Symbol
Ube2e2
Entrez Gene ID
218793
Gene Name
ubiquitin-conjugating enzyme E2E 2
Gene Aliases
BC016265
Chromosome Location
Chr. 14: 18573563 - 18894262 on Build GRCm38
UniGene ID
Mm.23551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144839.1 NP_659088.1 4 322
GenBank mRNA AK187401.1 - 2 122
BC016265.1 - 4 322 4923869

View Full Product Details