Gene Transcripts

Gene Symbol
Rtbdn
Entrez Gene ID
234542
Gene Name
retbindin
Gene Aliases
A330096I21Rik, BC022744, Rtbnd
Chromosome Location
Chr. 8: 84946263 - 84956606 on Build GRCm38
UniGene ID
Mm.461701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144929.2 NP_659178.1 6 1863
GenBank mRNA AK080715.1 - 6 1181
AY527275.1 - 6 1863
AY527276.1 - 7 1318
BC022744.1 - 5 1435 4527012
BC023051.1 - 5 1391 5361063
BC027079.1 - 5 1391 5357069

View Full Product Details