Gene Transcripts

Gene Symbol
Ube4a
Entrez Gene ID
140630
Gene Name
ubiquitination factor E4A
Gene Aliases
4732444G18Rik, 9930123J21Rik, UFD2b
Chromosome Location
Chr. 9: 44923127 - 44965600 on Build GRCm38
UniGene ID
Mm.215069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145400.3 NP_663375.3 5, 6 668
GenBank mRNA AK028721.1 - 5, 6 645
AK028850.1 - 5, 6 683
AK172893.1 - 5, 6 628
BC006649.1 - 6, 7 807 3498834
BC021406.1 - 5, 6 674
BC025043.1 - 5, 6 637

View Full Product Details