Gene Transcripts

Gene Symbol
Appl2
Entrez Gene ID
216190
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 2
Gene Aliases
Dip3b
Chromosome Location
Chr. 10: 83600033 - 83648854 on Build GRCm38
UniGene ID
Mm.282985
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145220.2 NP_660255.1 19, 20 1889
GenBank mRNA AK160075.1 - 19, 20 1868
AK160105.1 - 19, 20 1865
AK168403.1 - 19, 20 1852
AK168748.1 - 19, 20 1845
AK171512.1 - 19, 20 1845
AY113706.1 - 19, 20 1891
BC002232.1 - 7, 8 700
BC048906.1 - 19, 20 1827 6475576

View Full Product Details