Gene Transcripts

Gene Symbol
Klhl22
Entrez Gene ID
224023
Gene Name
kelch-like 22
Gene Aliases
2610318I18Rik, Kelchl
Chromosome Location
Chr. 16: 17759090 - 17793382 on Build GRCm38
UniGene ID
Mm.282807
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145479.4 NP_663454.3 3 382
GenBank mRNA AK028197.1 - 3 881
AK030669.1 - 3 327
AK086748.1 - 3 329
AK143514.1 - 3 327
AK204425.1 - 2 108
AK206875.1 - 2 108
AK214791.1 - 2 108
BC005800.1 - 3 360 2812589
CJ159851.1 - 3 374

View Full Product Details