Gene Transcripts

Gene Symbol
Erlin1
Entrez Gene ID
226144
Gene Name
ER lipid raft associated 1
Gene Aliases
2810439N09Rik, C80197, Keo4, Spfh1
Chromosome Location
Chr. 19: 44034943 - 44070493 on Build GRCm38
UniGene ID
Mm.279865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164359.1 NP_001157831.1 3 354
NM_001164360.1 NP_001157832.1 3 262
NM_145502.3 NP_663477.3 2 458
GenBank mRNA AK133557.1 - 2 201
AK145990.1 - 3 235
AK171750.1 - 1 1747
BC011220.1 - 3 226 4208238
BC021793.1 - 2 516
BC023849.1 - 2 321 5345801
CJ074662.1 - 3 263
CJ134812.1 - 3 335
CN702299.1 - 2 458

View Full Product Details