Gene Transcripts

Gene Symbol
Epb4.1l5
Entrez Gene ID
226352
Gene Name
erythrocyte protein band 4.1 like 5
Gene Aliases
1700030C16Rik, AL022914, BE37, E230025E14Rik, Epb41l5, Lulu1, NBL5
Chromosome Location
Chr. 1: 119545032 - 119649136 on Build GRCm38
UniGene ID
Mm.253156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145506.4 NP_663481.2 19, 20 1951
GenBank mRNA AK034676.1 - 19, 20 1873
AK044959.1 - 19, 20 1889
AK044996.1 - 19, 20 1897
AK047523.1 - 19, 20 1878
AK173198.1 - 19, 20 1909

View Full Product Details