Gene Transcripts

Gene Symbol
Adig
Entrez Gene ID
246747
Gene Name
adipogenin
Gene Aliases
SMAF-1, Smaf1
Chromosome Location
Chr. 2: 158502612 - 158508198 on Build GRCm38
UniGene ID
Mm.28800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145635.2 NP_663610.1 2 274
GenBank mRNA AK131610.1 - 2 224
AY092026.1 - 2 280
BC054059.1 - 2 222 3373967
BE624674.1 - 2 274
DQ223719.1 - 2 143

View Full Product Details