Gene Transcripts

Gene Symbol
Evc2
Entrez Gene ID
68525
Gene Name
Ellis van Creveld syndrome 2
Gene Aliases
1110017L09Rik, Lbn, limbin
Chromosome Location
Chr. 5: 37338478 - 37425055 on Build GRCm38
UniGene ID
Mm.25506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145920.3 NP_666032.1 2 286
GenBank mRNA AB083066.1 - 2 216
AK044977.1 - 2 157
AK134298.1 - 2 269
AK165051.1 - 2 171
BC037473.1 - 2 267 4237472
BC064473.1 - 2 277 3156380
BY270131.1 - 2 287

View Full Product Details