Gene Transcripts

Gene Symbol
Pank3
Entrez Gene ID
211347
Gene Name
pantothenate kinase 3
Gene Aliases
Chromosome Location
Chr. 11: 35769495 - 35791285 on Build GRCm38
UniGene ID
Mm.255044
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145962.2 NP_666074.1 2 541
GenBank mRNA AK039865.1 - 2 543
AK136305.1 - 2 231
BC027089.1 - 2 153 5355443
BC032188.1 - 2 587

View Full Product Details