Gene Transcripts

Gene Symbol
Gigyf2
Entrez Gene ID
227331
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
2610016F01Rik, A830080H02Rik, AI852361, AW259676, BC006835, Tnrc15, mKIAA0642
Chromosome Location
Chr. 1: 87326981 - 87450810 on Build GRCm38
UniGene ID
Mm.23065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110212.2 NP_001103682.1 24 3101
NM_146112.4 NP_666224.3 24 3119
GenBank mRNA AK122337.1 - 15 2190
AY176043.1 - 24 3065
BC027137.1 - 4 464
BC076610.1 - 24 3064
BC082811.1 - 5 575
BC089036.1 - 24 3075 30695300

View Full Product Details