Gene Transcripts

Gene Symbol
Gpr1
Entrez Gene ID
241070
Gene Name
G protein-coupled receptor 1
Gene Aliases
Chromosome Location
Chr. 1: 63182571 - 63223278 on Build GRCm38
UniGene ID
Mm.103354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146250.2 NP_666362.1 3 465
GenBank mRNA BC032934.1 - 3 393

View Full Product Details