Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
Chromosome Location
Chr. 3: 159988836 - 159996019 on Build GRCh38
UniGene ID
Hs.673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000882.3 NP_000873.2 7 1042
GenBank mRNA BC104982.1 - 7 974
BC104984.1 - 7 974 8143987
M65271.1 - 7 888
M65291.1 - 7 922

View Full Product Details