Gene Transcripts

Gene Symbol
Olfr168
Entrez Gene ID
258354
Gene Name
olfactory receptor 168
Gene Aliases
MOR271-1
Chromosome Location
Chr. 16: 19529980 - 19530918 on Build GRCm38
UniGene ID
Mm.377451
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146357.1 NP_666469.1 1 913
GenBank mRNA BC127969.1 - 1 917

View Full Product Details