Gene Transcripts

Gene Symbol
Olfr557
Entrez Gene ID
258358
Gene Name
olfactory receptor 557
Gene Aliases
MOR18-3
Chromosome Location
Chr. 7: 102698216 - 102699272 on Build GRCm38
UniGene ID
Mm.377455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146361.2 NP_666473.2 1 953
GenBank mRNA BC145899.1 - 1 953

View Full Product Details