Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000985.4 | NP_000976.1 | 4 | 480 | |
NM_001035006.2 | NP_001030178.1 | 4 | 231 | ||
NM_001199340.1 | NP_001186269.1 | 5 | 329 | ||
NM_001199341.1 | NP_001186270.1 | 4 | 330 | ||
NM_001199342.1 | NP_001186271.1 | 5 | 428 | ||
NM_001199343.1 | NP_001186272.1 | 5 | 411 | ||
NM_001199344.1 | NP_001186273.1 | 4 | 313 | ||
NM_001199345.1 | NP_001186274.1 | 3 | 190 | ||
GenBank mRNA | AK304659.1 | - | 3 | 104 | |
AK311720.1 | - | 2 | 881 | ||
AK311828.1 | - | 4 | 145 | ||
BC000502.2 | - | 3 | 383 | 2821464 | |
BC017831.1 | - | 1 | 149 | 4251433 | |
BC066323.1 | - | 1 | 145 | 4797583 | |
BC066324.1 | - | 1 | 149 | 4799029 | |
BC106031.1 | - | 1 | 116 | 4540700 | |
BF670086.1 | - | 327 | |||
BG497914.1 | - | 4 | 396 | ||
BG758380.1 | - | 5 | 347 | ||
BG827384.1 | - | 4 | 231 | ||
BI086293.1 | - | 4 | 202 | ||
BQ877039.1 | - | 5 | 369 | ||
BQ882430.1 | - | 4 | 296 | ||
CB178432.1 | - | 3 | 396 | ||
CD386294.1 | - | 4 | 250 | ||
CD387752.1 | - | 4 | 335 | ||
X53777.1 | - | 3 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199355.1 | NP_001186284.1 | 3 | 123 | |
NM_001199356.1 | NP_001186285.1 | 3 | 190 | ||
GenBank mRNA | BI461805.1 | - | 3 | 109 | |
BX393840.2 | - | 3 | 125 |