Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289722.1 | NP_001276651.1 | 3 | 310 | |
NM_001289723.1 | NP_001276652.1 | 3 | 275 | ||
NM_153138.4 | NP_694778.1 | 3 | 305 | ||
GenBank mRNA | AJ437262.1 | - | 2 | 106 | |
AK031046.1 | - | 3 | 272 | ||
AK156547.1 | - | 3 | 310 | ||
BC049788.1 | - | 3 | 228 | 6514406 | |
BY200699.1 | - | 3 | 276 | ||
BY252790.1 | - | 3 | 306 | ||
BY285630.1 | - | 3 | 265 |