Gene Transcripts

Gene Symbol
Klhl15
Entrez Gene ID
236904
Gene Name
kelch-like 15
Gene Aliases
6330500C13Rik
Chromosome Location
Chr. X: 94234495 - 94277724 on Build GRCm38
UniGene ID
Mm.115533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039059.1 NP_001034148.1 3 489
NM_001039060.1 NP_001034149.1 4 540
NM_001039061.1 NP_001034150.1 2 584
NM_153165.2 NP_694805.1 4 540
GenBank mRNA AK078112.1 - 2 525
AK081546.1 - 3 493
AK133055.1 - 3 254
AK169500.1 - 4 483
BC036978.1 - 4 477
BC141165.1 - 3 254 9055777

View Full Product Details