Gene Transcripts

Gene Symbol
Ints9
Entrez Gene ID
210925
Gene Name
integrator complex subunit 9
Gene Aliases
BC028953, D14Ertd231e
Chromosome Location
Chr. 14: 64950045 - 65039835 on Build GRCm38
UniGene ID
Mm.71332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253731.1 NP_001240660.1 3, 4 552
NM_153414.4 NP_700463.2 3, 4 520
GenBank mRNA AK035769.1 - 3, 4 264
AK038979.1 - 3, 4 253
AK077634.1 - 3, 4 521
AK136992.1 - 3, 4 247
AK150195.1 - 3, 4 247
BC028953.1 - 2, 3 210 5362016
BC055700.1 - 3, 4 219 6808816
BY195822.1 - 3, 4 285

View Full Product Details