Gene Transcripts

Gene Symbol
Dis3l2
Entrez Gene ID
208718
Gene Name
DIS3 mitotic control homolog (S. cerevisiae)-like 2
Gene Aliases
4930429A22Rik, 8030493P09Rik
Chromosome Location
Chr. 1: 86703779 - 87050097 on Build GRCm38
UniGene ID
Mm.389152
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172157.1 NP_001165628.1 3 302
NM_153530.2 NP_705758.1 3 302
GenBank mRNA AK029652.1 - 3 279
AK031180.1 - 3 287
AK040613.1 - 3 279
AK079867.1 - 3 299
BC036177.1 - 3 263 5004341
CJ063044.1 - 3 281

View Full Product Details