Gene Transcripts

Gene Symbol
Ttc41
Entrez Gene ID
103220
Gene Name
tetratricopeptide repeat domain 41
Gene Aliases
AI449705, AU041509, Gnn
Chromosome Location
Chr. 10: 86705632 - 86776844 on Build GRCm38
UniGene ID
Mm.440762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003910.2 NP_001003910.2 2 784
NM_001003939.2 NP_001003939.2 3 893
NM_153595.3 NP_705823.3 1 577
GenBank mRNA AK133195.1 - 2 724
AY651019.1 - 4 976
AY651020.1 - 1 424
AY651021.1 - 1 424
BC030307.1 - 3 871

View Full Product Details