Gene Transcripts

Gene Symbol
Cpne7
Entrez Gene ID
102278
Gene Name
copine VII
Gene Aliases
AW047065
Chromosome Location
Chr. 8: 123117315 - 123135185 on Build GRCm38
UniGene ID
Mm.36434
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170684.2 NP_733785.1 13, 14 1424
GenBank mRNA AK049823.1 - 10, 11 2566
AK162900.1 - 12, 13 2043
BC119328.1 - 13, 14 1323 8734077
BC137595.1 - 13, 14 1323 8860614
BM946575.1 - 6, 7 487

View Full Product Details