Gene Transcripts

Gene Symbol
Rufy1
Entrez Gene ID
216724
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
3000002E04Rik, Rabip4, ZFYVE12
Chromosome Location
Chr. 11: 50389286 - 50431111 on Build GRCm38
UniGene ID
Mm.390400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172557.2 NP_766145.1 18 2282
GenBank mRNA AK046633.1 - 18 2283

View Full Product Details