Gene Transcripts

Gene Symbol
Dpp9
Entrez Gene ID
224897
Gene Name
dipeptidylpeptidase 9
Gene Aliases
6430584G11Rik, A330078I11, DPP IX, DPRP2
Chromosome Location
Chr. 17: 56186674 - 56218889 on Build GRCm38
UniGene ID
Mm.205535
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172624.3 NP_766212.2 7, 8 1034
GenBank mRNA AK039652.1 - 6, 7 1127
AK050021.1 - 7, 8 976
AK050023.1 - 7, 8 1036
AK078301.1 - 7, 8 982
AK131178.1 - 4, 5 2897
AK219044.1 - 1, 2 51
BC057631.1 - 7, 8 1033 4191886

View Full Product Details