Gene Transcripts

Gene Symbol
Nfrkb
Entrez Gene ID
235134
Gene Name
nuclear factor related to kappa B binding protein
Gene Aliases
A530090G11Rik
Chromosome Location
Chr. 9: 31386161 - 31421333 on Build GRCm38
UniGene ID
Mm.238146
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172766.3 NP_766354.2 16 1808
GenBank mRNA AK041206.1 - 16 2046
AK049804.1 - 2 183
BC033595.1 - 16 1798 4947357
BC059861.1 - 16 1855
BC059900.1 - 7 1455
BC060076.1 - 16 2037

View Full Product Details