Gene Transcripts

Gene Symbol
Slc9a6
Entrez Gene ID
236794
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 6
Gene Aliases
3732426M05, 6430520C02Rik, NHE6, mKIAA0267
Chromosome Location
Chr. X: 56609557 - 56664230 on Build GRCm38
UniGene ID
Mm.17815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172780.3 NP_766368.2 16 2296
GenBank mRNA AK028350.1 - 2 555
AK032326.1 - 16 2277

View Full Product Details