Gene Transcripts

Gene Symbol
Dip2b
Entrez Gene ID
239667
Gene Name
DIP2 disco-interacting protein 2 homolog B (Drosophila)
Gene Aliases
4932422C22, AI317237, AI854602, mKIAA1463
Chromosome Location
Chr. 15: 100038318 - 100219474 on Build GRCm38
UniGene ID
Mm.243658
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159361.1 NP_001152833.1 7 961
NM_172819.3 NP_766407.2 2 176
GenBank mRNA AK030064.1 - 2 177
AK147562.1 - 7 897
BC137857.1 - 2 163 8860876
BC137858.1 - 2 163 8860877

View Full Product Details