Gene Transcripts

Gene Symbol
Lrp12
Entrez Gene ID
239393
Gene Name
low density lipoprotein-related protein 12
Gene Aliases
AI848829, C820005L12Rik, LRP-12
Chromosome Location
Chr. 15: 39870602 - 39944009 on Build GRCm38
UniGene ID
Mm.39874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172814.3 NP_766402.1 5 1297
GenBank mRNA AK050512.1 - 5 1298
AK084675.1 - 1 97
AK213199.1 - 1 236
BC058345.1 - 4 1477 6822657

View Full Product Details