Gene Transcripts

Gene Symbol
Fsd2
Entrez Gene ID
244091
Gene Name
fibronectin type III and SPRY domain containing 2
Gene Aliases
9830160G03Rik, Spryd1
Chromosome Location
Chr. 7: 81534354 - 81566981 on Build GRCm38
UniGene ID
Mm.38773
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172904.2 NP_766492.2 2 668
GenBank mRNA AK028142.1 - 2 660
AK036674.1 - 634

View Full Product Details