Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173413.3 | NP_775589.1 | 3, 4 | 318 | |
GenBank mRNA | AB232601.1 | - | 3, 4 | 246 | |
AK051218.1 | - | 3, 4 | 268 | ||
AK084650.1 | - | 3, 4 | 268 | ||
AK155621.1 | - | 3, 4 | 268 | ||
BC059208.1 | - | 3, 4 | 300 | 6405462 | |
BC157974.1 | - | 3, 4 | 266 | 9007296 | |
BY322741.1 | - | 3, 4 | 319 |