Gene Transcripts

Gene Symbol
Cyld
Entrez Gene ID
74256
Gene Name
cylindromatosis (turban tumor syndrome)
Gene Aliases
2010013M14Rik, 2900009M21Rik, C130039D01Rik, CDMT, CYLD1, EAC, mKIAA0849
Chromosome Location
Chr. 8: 88697001 - 88751946 on Build GRCm38
UniGene ID
Mm.490395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128170.2 NP_001121642.1 18 3471
NM_001128171.2 NP_001121643.1 18 3596
NM_001276279.1 NP_001263208.1 16 3047
NM_173369.3 NP_775545.1 17 3462
GenBank mRNA AK008239.1 - 1 336
AK122389.1 - 18 3577
AK133479.1 - 17 3426
AK134926.1 - 5 1154
AK147204.1 - 18 3439
AK147426.1 - 18 3434
AK167424.1 - 16 3011
BC023702.1 - 2 720
BC024596.1 - 5 1140
BC042438.1 - 17 3415 3983771
BC049879.1 - 8 1324

View Full Product Details