Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173413.3 | NP_775589.1 | 6, 7 | 560 | |
GenBank mRNA | AB232601.1 | - | 6, 7 | 488 | |
AK031251.1 | - | 6, 7 | 507 | ||
AK084650.1 | - | 6, 7 | 510 | ||
AK086085.1 | - | 1, 2 | 102 | ||
AK155621.1 | - | 6, 7 | 510 | ||
BC059208.1 | - | 6, 7 | 542 | 6405462 | |
BC157974.1 | - | 6, 7 | 508 | 9007296 |