Gene Transcripts

Gene Symbol
Lrrc28
Entrez Gene ID
67867
Gene Name
leucine rich repeat containing 28
Gene Aliases
1300004K21Rik, 2210012C09Rik, 2310058O11Rik
Chromosome Location
Chr. 7: 67513410 - 67645331 on Build GRCm38
UniGene ID
Mm.31247
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027413.1 NP_081689.1 2 270
NM_175124.5 NP_780333.3 2 270
GenBank mRNA AI663251.1 - 2 270
AK004904.1 - 2 239
AK008713.1 - 2 238
AK050154.1 - 2 242
AK077270.1 - 2 266
AK159417.1 - 2 239
AK183719.1 - 1 161
BC052051.1 - 1 177
BC086694.1 - 2 161 30697866

View Full Product Details