Gene Transcripts

Gene Symbol
Ankrd46
Entrez Gene ID
68839
Gene Name
ankyrin repeat domain 46
Gene Aliases
1110054N06Rik, AI314978, AI987733
Chromosome Location
Chr. 15: 36477668 - 36503426 on Build GRCm38
UniGene ID
Mm.396537
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175134.4 NP_780343.1 5 1622
GenBank mRNA AB076382.1 - 5 1617
AK004254.1 - 1 629
AK079270.1 - 3 1181
AK088304.1 - 5 1583
AK148049.1 - 5 1567
BC035553.1 - 5 3200
BC086659.1 - 5 1589 30607272
BC094595.1 - 5 1585 5707829

View Full Product Details