Gene Transcripts

Gene Symbol
Bola3
Entrez Gene ID
78653
Gene Name
bolA-like 3 (E. coli)
Gene Aliases
1810056O20Rik
Chromosome Location
Chr. 6: 83349484 - 83358392 on Build GRCm38
UniGene ID
Mm.391470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175277.4 NP_780486.1 2 148
GenBank mRNA AK028102.1 - 2 149
AK131950.1 - 157
AK160488.1 - 2 152
AK213244.1 - 1 83
BC049748.1 - 2 168
BC116947.1 - 2 186 40126266
BC116949.1 - 2 186 40126268

View Full Product Details